Learn More
New England Biolabs, Inc. EarI – 500 units
SDP
Catalog No. 50812188
Encompass
A restriction endonuclease that recognizes the sequence CTCTTCN^NNN_.
Catalog No. 50-812-188 Supplier New England Biolabs, Inc. Supplier No. R0528S
A restriction endonuclease that recognizes the sequence CTCTTCN^NNN_.