Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Earle's Balanced Salts without Sodium Bicarbonate

Catalog No. MP91700122 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP91700122 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP91700122 Supplier MP Biomedicals™ Supplier No. 091700122
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For mammalian cell culture

TRUSTED_SUSTAINABILITY

Specifications

Form Powder
For Use With (Application) Mammalian Cell Culture
Quantity 10 L
Formulation Component Inorganic Salts Calcium Chloride [CaCl2 · 2H2O] Dihydrate 264.90000mg/L; Magnesium Sulfate [MgSO4] 97.70000mg/L; Potassium Chloride [KCl] 400.00000mg/L; Sodium Chloride [NaCl] 6800.00000mg/L; Sodium Phosphate Monobasic [NaH2PO4 · H2O] Monohydrate 140.00000mg/L; Other Dextrose 1000.00000mg/L; Phenol Red Sodium Salt 17.00000mg/L
Content And Storage Store at 4°C
Description Earle's Balanced Salts
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.