Learn More
Sigma Aldrich Fine Chemicals Biosciences Earle’s Balanced Salts | 6x500mL
Catalog No. NC1578258
Encompass
Phenol red has been shown to interfere with the growth of some cells at cloning densities. Use this medium when working with stem cells or when growing cells at low densities.
Catalog No. NC1578258 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. E30246X500ML
Phenol red has been shown to interfere with the growth of some cells at cloning densities. Use this medium when working with stem cells or when growing cells at low densities.