Learn More
Electron Microscopy Sciences Earl's Balanced Salts Formula III 500 ML
SDP
Catalog No. 50980251
Encompass_Preferred
All of our Earle's Balanced Salt Solution contains: Potassium Chloride, Sodium Chloride, Sodium Phosphate Monobasic, and D-glucose. Some formulations may contain additional components. Use the table below as a quick reference for identifying the one that best suits your application. All of the solutions are prepared 1X strength.
Catalog No. 50-980-251 Supplier Electron Microscopy Sciences Supplier No. 1176705
All of our Earle's Balanced Salt Solution contains: Potassium Chloride, Sodium Chloride, Sodium Phosphate Monobasic, and D-glucose. Some formulations may contain additional components. Use the table below as a quick reference for identifying the one that best suits your application. All of the solutions are prepared 1X strength.