Learn More
Cayman Chemical Early ApoptosIs DetectIon
SDP
Catalog No. 502892361
Encompass_Preferred
Cayman's EArly Apoptosis Detection Assay Kit employs FITC-conjugated annexin V as a probe for phosphatidylserine (PS) on the outer membrane of apoptotic cells, TMRE as a probe for ΔΨm, DAPI as an indicator of membrane permeability/cell viability, and TO-PRO-3 to indicate the selective permeability of pannexin channels. The EArly Apoptosis Detection Assay Kit allows phenotypic characterization of multiple different cell death parameters at the single-cell level.
Catalog No. 50-289-2361 Supplier Cayman Chemical Supplier No. 601360100 TEST
Cayman's EArly Apoptosis Detection Assay Kit employs FITC-conjugated annexin V as a probe for phosphatidylserine (PS) on the outer membrane of apoptotic cells, TMRE as a probe for ΔΨm, DAPI as an indicator of membrane permeability/cell viability, and TO-PRO-3 to indicate the selective permeability of pannexin channels. The EArly Apoptosis Detection Assay Kit allows phenotypic characterization of multiple different cell death parameters at the single-cell level.