Learn More
ORS Nasco EARPLG REUS RG HEARPK 100PR/PK
Catalog No. 502551617
Encompass_Preferred
Honeywell part# FUS30-HP. Honeywell Howard Leight Fusion Ear Plugs Size: Regular Corded: Corded Disposable: Reusable Includes: Pocket-size case Metal Detectable: No Noise Reduction Rating: 27 dB Plug Shape: Flanged ANSI S3.1974: Yes Cord Color: Blue yellow Cord Material: Nylon CSA Z94.2: Class AL HearPack case
Catalog No. 50-255-1617 Supplier ORS Nasco Supplier No. 154FUS30HP
Honeywell part# FUS30-HP. Honeywell Howard Leight Fusion Ear Plugs Size: Regular Corded: Corded Disposable: Reusable Includes: Pocket-size case Metal Detectable: No Noise Reduction Rating: 27 dB Plug Shape: Flanged ANSI S3.1974: Yes Cord Color: Blue yellow Cord Material: Nylon CSA Z94.2: Class AL HearPack case