Learn More
MSC Earplug Dispenser Refill, 3M, Foam, 29 dB NRR, 500 Pr
Catalog No. 5000687718
Encompass
This earplug dispenser refill contains 500 pairs of foam, barrel-shaped, disposable earplugs. They are yellow and packaged in a bottle.
  • Earplug refill type
  • Foam plug material
  • Barrel plug shape
  • Noise reduction rating of 29 dB
  • One touch dispensing method
  • Disposable
  • Bottle container type
  • Package quantity of 500 pairs
  • Yellow plug color
  • Universal plug size
  • Uncorded style
  • Flame-resistant and moisture-resistant
  • Compatible with validation system
  • CSA Class AL
Catalog No. 50-006-87718 Supplier MSC Supplier No. 02948610
This earplug dispenser refill contains 500 pairs of foam, barrel-shaped, disposable earplugs. They are yellow and packaged in a bottle.
  • Earplug refill type
  • Foam plug material
  • Barrel plug shape
  • Noise reduction rating of 29 dB
  • One touch dispensing method
  • Disposable
  • Bottle container type
  • Package quantity of 500 pairs
  • Yellow plug color
  • Universal plug size
  • Uncorded style
  • Flame-resistant and moisture-resistant
  • Compatible with validation system
  • CSA Class AL