Learn More
ORS Nasco EARPLUG MAX LITE CORD 100PR/PK
Catalog No. 502551611
Encompass_Preferred
Honeywell part# LPF-30. Honeywell Max-Lite Ear Plugs Corded: Corded Cord Material: Polycarbonate Poly bag Disposable: Yes Metal Detectable: No Noise Reduction Rating: 30 dB Plug Shape: T-Shape Size: Small ANSI S3.1974: Yes Cord Color: Yellow CSA Z94.2: Class AL High Visibility: No
Catalog No. 50-255-1611 Supplier ORS Nasco Supplier No. 154LPF30
Honeywell part# LPF-30. Honeywell Max-Lite Ear Plugs Corded: Corded Cord Material: Polycarbonate Poly bag Disposable: Yes Metal Detectable: No Noise Reduction Rating: 30 dB Plug Shape: T-Shape Size: Small ANSI S3.1974: Yes Cord Color: Yellow CSA Z94.2: Class AL High Visibility: No