Learn More
ORS Nasco EARPLUG MAX1 W/O CORD 200PR/PK
Catalog No. 502551610
Encompass_Preferred
Honeywell part# MAX-1. Honeywell Max Ear Plugs Corded: Uncorded Pairs per Package: 200 Pairs/Pk. Disposable: Yes Metal Detectable: No Noise Reduction Rating: 33 dB Plug Shape: Bell Size: Universal ANSI S3.1974: Yes CSA Z94.2: Class AL High Visibility: Yes Poly bag Plug Color: Orange Plug Material: Foam
Catalog No. 50-255-1610 Supplier ORS Nasco Supplier No. 154MAX1
Honeywell part# MAX-1. Honeywell Max Ear Plugs Corded: Uncorded Pairs per Package: 200 Pairs/Pk. Disposable: Yes Metal Detectable: No Noise Reduction Rating: 33 dB Plug Shape: Bell Size: Universal ANSI S3.1974: Yes CSA Z94.2: Class AL High Visibility: Yes Poly bag Plug Color: Orange Plug Material: Foam