Learn More
MSC Howard Leight Earplugs, 26 dB NRR, Vinyl, Corded, Push-In, 100/Pk.
Catalog No. 502370324
Encompass
These reusable earplugs are designed with a bell shape and a corded design, made from vinyl, and feature a push-in insertion method. They are non-metal detectable and come in a universal size, offering convenient hearing protection.
  • Reusable
  • Corded design
  • Push-in insertion method
  • Non-metal detectable
  • Universal size
  • Noise Reduction Rating of 26 dB (NRR)
Catalog No. 50-237-0324 Supplier MSC Supplier No. 09798158
These reusable earplugs are designed with a bell shape and a corded design, made from vinyl, and feature a push-in insertion method. They are non-metal detectable and come in a universal size, offering convenient hearing protection.
  • Reusable
  • Corded design
  • Push-in insertion method
  • Non-metal detectable
  • Universal size
  • Noise Reduction Rating of 26 dB (NRR)