Learn More
MSC Howard Leight Earplugs, 25 dB NRR, Flanged, Corded, 100 Pairs
Catalog No. 502370317
Encompass
Howard Leight SmartFit earplugs offer a personalized fit by utilizing patented conforming material technology that adapts to the wearer's ear canal using body heat. This design ensures superior comfort and simplifies inventory management as a single product fits most users.
  • Patented Conforming Material Technology adapts to ear canal using body heat.
  • Delivers superior comfort and a truly individual fit.
  • Simplifies inventory control as a single product fits almost every wearer.
  • Detachable cord system and HearPack storage case included.
  • Made from thermoplastic elastomer (TPE).
  • Features a 25 dB Noise Reduction Rating (NRR).
  • Designed with a 3-flange shape for effective sealing.
  • Reusable for extended use.
Catalog No. 50-237-0317 Supplier MSC Supplier No. 05839048
Howard Leight SmartFit earplugs offer a personalized fit by utilizing patented conforming material technology that adapts to the wearer's ear canal using body heat. This design ensures superior comfort and simplifies inventory management as a single product fits most users.
  • Patented Conforming Material Technology adapts to ear canal using body heat.
  • Delivers superior comfort and a truly individual fit.
  • Simplifies inventory control as a single product fits almost every wearer.
  • Detachable cord system and HearPack storage case included.
  • Made from thermoplastic elastomer (TPE).
  • Features a 25 dB Noise Reduction Rating (NRR).
  • Designed with a 3-flange shape for effective sealing.
  • Reusable for extended use.