Learn More
Essendant EARPLUGPELTOR OPTIME 105
Supplier:  Essendant MMMH10AHV
Encompass_Preferred
Twin cup design minimizes resonance thus achieving maximum high and low frequency attenuation Pivoting earcups for optimum fit Padded steel headband with four point suspension for extra comfort Material(s)- ABS Plastic Ear Cushion Material- Liquid/Foam Noise Reduction Rate (NRR)- 30 dB Ear Muff Style- Over-The-Head
Catalog No. 50-000-35844