Learn More
MSC EARPLUGS
Catalog No. 502834680
Encompass
These premolded, triple flange earplugs are designed for comfortable fit and effective noise reduction in various ear sizes. They are washable and reusable, offering a cost-effective and hygienic hearing protection solution.
  • Premolded, triple flange design fits comfortably
  • Accommodates a wide range of ear sizes
  • No need to roll before fitting, keeping tips clean
  • Soft polymer is washable and reusable
  • Reduces waste and costs
  • Poly bag keeps each pair accessible for reuse
  • Noise Reduction Rating (NRR) of 25 dB
  • CSA Class AL rated
  • Test compatible with E-A-Rfit Validation System
Catalog No. 50-283-4680 Supplier MSC Supplier No. 06528012
These premolded, triple flange earplugs are designed for comfortable fit and effective noise reduction in various ear sizes. They are washable and reusable, offering a cost-effective and hygienic hearing protection solution.
  • Premolded, triple flange design fits comfortably
  • Accommodates a wide range of ear sizes
  • No need to roll before fitting, keeping tips clean
  • Soft polymer is washable and reusable
  • Reduces waste and costs
  • Poly bag keeps each pair accessible for reuse
  • Noise Reduction Rating (NRR) of 25 dB
  • CSA Class AL rated
  • Test compatible with E-A-Rfit Validation System