Learn More
MSC EARPLUGS
Catalog No. 502834654
Encompass
These earplugs are designed for hearing protection with a 29 dB noise reduction rating. They are reusable and feature a bell shape for comfortable, easy insertion without needing to be rolled down, promoting hygiene.
  • Reusable
  • Corded attachment style
  • Bell-shaped plug for comfort
  • Noise reduction rating of 29 dB
  • Foam material
  • Roll down insertion method
  • Universal size
  • Non-metal detectable
  • Yellow vinyl cord
  • No-touch series
  • Meets CSA Class AL standards
Catalog No. 50-283-4654 Supplier MSC Supplier No. 93071322
These earplugs are designed for hearing protection with a 29 dB noise reduction rating. They are reusable and feature a bell shape for comfortable, easy insertion without needing to be rolled down, promoting hygiene.
  • Reusable
  • Corded attachment style
  • Bell-shaped plug for comfort
  • Noise reduction rating of 29 dB
  • Foam material
  • Roll down insertion method
  • Universal size
  • Non-metal detectable
  • Yellow vinyl cord
  • No-touch series
  • Meets CSA Class AL standards