Learn More
MSC 3M Earplugs, 33 dB, Foam, Bullet, Corded
Catalog No. 502834906
Encompass
The E-A-Rsoft Yellow Neons earplugs are designed to reduce exposure to hazardous noise. Made from soft, pliable foam with a bullet shape and corded design, they offer comfort and protection with a Noise Reduction Rating (NRR) of 33 dB. These reusable earplugs are suitable for various industrial and laboratory applications.
  • Tapered shape and smooth soft texture for enhanced fit and comfort
  • Slow-recovery foam allows for easier insertion
  • Compatible with the E-A-Rfit validation system
  • High visibility neon yellow coloring
  • Testable to validate protection level for individual users
  • Soft, pliable foam for extended wear comfort
  • Low-pressure foam reduces noticeability
  • Easy roll-down for quick insertions
  • Poly bag maintains cleanliness before use
  • Regular size fits most ear canals
  • CSA Class AL certified
Catalog No. 50-283-4906 Supplier MSC Supplier No. 08353179
The E-A-Rsoft Yellow Neons earplugs are designed to reduce exposure to hazardous noise. Made from soft, pliable foam with a bullet shape and corded design, they offer comfort and protection with a Noise Reduction Rating (NRR) of 33 dB. These reusable earplugs are suitable for various industrial and laboratory applications.
  • Tapered shape and smooth soft texture for enhanced fit and comfort
  • Slow-recovery foam allows for easier insertion
  • Compatible with the E-A-Rfit validation system
  • High visibility neon yellow coloring
  • Testable to validate protection level for individual users
  • Soft, pliable foam for extended wear comfort
  • Low-pressure foam reduces noticeability
  • Easy roll-down for quick insertions
  • Poly bag maintains cleanliness before use
  • Regular size fits most ear canals
  • CSA Class AL certified