Learn More
MSC Jackson Safety Earplugs, 26 dB NRR, Rubber, Metal-Detectable, 100 Pairs
Catalog No. 502834616
Encompass
These earplugs offer a 26 dB Noise Reduction Rating, featuring a rubber, flanged, and push-in design. They are corded and metal-detectable for enhanced safety and convenience, supplied in packs of 100 reusable pairs.
  • Flanged design for comfortable fit
  • Push-in insertion method
  • Corded for convenience and to prevent loss
  • Metal-detectable for use in sensitive environments
  • Reusable for extended use
  • Offers a 26 dB Noise Reduction Rating
  • Constructed from durable rubber material
  • Universal size to fit most users
  • Complies with S3.19-1974 standards
Catalog No. 50-283-4616 Supplier MSC Supplier No. 57681843
These earplugs offer a 26 dB Noise Reduction Rating, featuring a rubber, flanged, and push-in design. They are corded and metal-detectable for enhanced safety and convenience, supplied in packs of 100 reusable pairs.
  • Flanged design for comfortable fit
  • Push-in insertion method
  • Corded for convenience and to prevent loss
  • Metal-detectable for use in sensitive environments
  • Reusable for extended use
  • Offers a 26 dB Noise Reduction Rating
  • Constructed from durable rubber material
  • Universal size to fit most users
  • Complies with S3.19-1974 standards