Learn More
MSC Moldex Earplugs, 30 dB, Foam, Corded, Non-Metal Detectable, 100 Pairs
Catalog No. 502834621
Encompass
These earplugs provide effective hearing protection, featuring a 30 dB Noise Reduction Rating for use in loud environments. These disposable, corded earplugs are designed for easy push-in insertion and are suitable for a universal fit.
  • Disposable design for single-use applications
  • Provides 30 dB Noise Reduction Rating (NRR) ensuring significant sound attenuation
  • Constructed from soft foam material for comfort
  • Bell-shaped plug design for secure and comfortable fit
  • Push-in insertion method for easy and quick application
  • Equipped with a durable polyurethane cord to prevent loss
  • Universal size designed to fit most users
  • Non-metal detectable construction
Catalog No. 50-283-4621 Supplier MSC Supplier No. 61490553
These earplugs provide effective hearing protection, featuring a 30 dB Noise Reduction Rating for use in loud environments. These disposable, corded earplugs are designed for easy push-in insertion and are suitable for a universal fit.
  • Disposable design for single-use applications
  • Provides 30 dB Noise Reduction Rating (NRR) ensuring significant sound attenuation
  • Constructed from soft foam material for comfort
  • Bell-shaped plug design for secure and comfortable fit
  • Push-in insertion method for easy and quick application
  • Equipped with a durable polyurethane cord to prevent loss
  • Universal size designed to fit most users
  • Non-metal detectable construction