Learn More
MSC 3M Earplugs, Foam, Uncorded, 33 dB NRR, Yellow
Catalog No. 502835093
Encompass
The 3M E-A-Rsoft Yellow Neons Earplugs are engineered for industrial and occupational environments, offering effective hearing protection. They feature a tapered, bullet shape and smooth, slow-recovery polyurethane foam for easy insertion and a comfortable, secure fit.
  • Tapered shape and smooth soft texture for fit and comfort.
  • Slow-recovery foam designed for easier insertion.
  • Compatible with the E-A-Rfit Validation System.
  • High visibility with neon yellow coloring to verify compliance.
  • Provides 33 dB noise reduction rating.
  • Made from polyurethane foam.
  • Available in universal size.
  • Reusable.
Catalog No. 50-283-5093 Supplier MSC Supplier No. 08353138
Add to Cart
Add to Cart
The 3M E-A-Rsoft Yellow Neons Earplugs are engineered for industrial and occupational environments, offering effective hearing protection. They feature a tapered, bullet shape and smooth, slow-recovery polyurethane foam for easy insertion and a comfortable, secure fit.
  • Tapered shape and smooth soft texture for fit and comfort.
  • Slow-recovery foam designed for easier insertion.
  • Compatible with the E-A-Rfit Validation System.
  • High visibility with neon yellow coloring to verify compliance.
  • Provides 33 dB noise reduction rating.
  • Made from polyurethane foam.
  • Available in universal size.
  • Reusable.