Learn More
MSC 3M Earplugs, 33 dB NRR, Foam, Corded, Neon Yellow
Catalog No. 502835094
Encompass
These earplugs are designed to seal the ear canal with soft, slow-recovery foam, providing noise reduction and improved comfort. The slow-recovery foam facilitates easier insertion, allowing time for proper placement before full expansion. Featuring a high-visibility neon yellow coloring, these reusable earplugs are easy to spot for compliance verification.
  • Reduces noise with a 33 dB Noise Reduction Rating
  • Made from soft, slow-recovery foam for comfort
  • Features a corded design
  • High visibility neon yellow color
  • Reusable for extended use
  • Bullet shape for easy insertion
  • Universal size fits most ear canals
  • Non-metal detectable
Catalog No. 50-283-5094 Supplier MSC Supplier No. 08353195
These earplugs are designed to seal the ear canal with soft, slow-recovery foam, providing noise reduction and improved comfort. The slow-recovery foam facilitates easier insertion, allowing time for proper placement before full expansion. Featuring a high-visibility neon yellow coloring, these reusable earplugs are easy to spot for compliance verification.
  • Reduces noise with a 33 dB Noise Reduction Rating
  • Made from soft, slow-recovery foam for comfort
  • Features a corded design
  • High visibility neon yellow color
  • Reusable for extended use
  • Bullet shape for easy insertion
  • Universal size fits most ear canals
  • Non-metal detectable