Learn More
MSC Moldex Earplugs, NRR 33 dB, Foam, Corded, Disposable
Catalog No. 502835130
Encompass
These earplugs provide high-level hearing protection with a Noise Reduction Rating of 33 dB. They are designed with a unique contoured shape and soft, slow-recovery foam to ensure a comfortable and secure fit for various ear canal sizes. The corded, disposable design makes them convenient for use in demanding environments.
  • 100% PVC-free material.
  • Highest independently tested NRR 33.
  • Exclusive contoured shape for a secure fit.
  • Soft, slow-recovery foam for maximum comfort.
  • Non-irritating, smooth surface.
  • Bright green color for easy compliance checks.
  • Independently tested to ANSI S3.19-1974.
Catalog No. 50-283-5130 Supplier MSC Supplier No. 68041276
Add to Cart
Add to Cart
These earplugs provide high-level hearing protection with a Noise Reduction Rating of 33 dB. They are designed with a unique contoured shape and soft, slow-recovery foam to ensure a comfortable and secure fit for various ear canal sizes. The corded, disposable design makes them convenient for use in demanding environments.
  • 100% PVC-free material.
  • Highest independently tested NRR 33.
  • Exclusive contoured shape for a secure fit.
  • Soft, slow-recovery foam for maximum comfort.
  • Non-irritating, smooth surface.
  • Bright green color for easy compliance checks.
  • Independently tested to ANSI S3.19-1974.