Learn More
MSC Moldex Earplug, NRR 27 dB, Corded, Flanged, Reusable
Catalog No. 502835336
Encompass
Moldex Rockets Corded Reusable Earplugs are designed for hearing protection in demanding work environments. They feature a soft triple-flange design with an air bubble in the tip for cushioned comfort and a superior seal. These earplugs are washable and reusable, offering an NRR of 27 dB. Each pair includes a Pocket-Pak carrying case for hygienic storage between uses, providing a cost-effective and sustainable solution that is 100% PVC-Free and Made in America.
  • Air-cushioned for comfort with an air bubble in the tip
  • Easy grip handle for simple insertion into the ear
  • Washable and reusable soft triple-flange plug
  • Pocket-Pak carrying case for hygienic storage between uses
  • High NRR 27 dB, independently tested for protection
  • Made in America, ensuring quality
  • 100% PVC-Free for sustainable solutions
Catalog No. 50-283-5336 Supplier MSC Supplier No. 00412643
Add to Cart
Add to Cart
Moldex Rockets Corded Reusable Earplugs are designed for hearing protection in demanding work environments. They feature a soft triple-flange design with an air bubble in the tip for cushioned comfort and a superior seal. These earplugs are washable and reusable, offering an NRR of 27 dB. Each pair includes a Pocket-Pak carrying case for hygienic storage between uses, providing a cost-effective and sustainable solution that is 100% PVC-Free and Made in America.
  • Air-cushioned for comfort with an air bubble in the tip
  • Easy grip handle for simple insertion into the ear
  • Washable and reusable soft triple-flange plug
  • Pocket-Pak carrying case for hygienic storage between uses
  • High NRR 27 dB, independently tested for protection
  • Made in America, ensuring quality
  • 100% PVC-Free for sustainable solutions