Learn More
MSC Cordova Earplugs, 27 dB NRR, Thermoplastic Rubber, Flanged, Push-In, Corded
Catalog No. 502835483
Encompass
Cordova earplugs provide effective hearing protection with a 27 dB Noise Reduction Rating. Constructed from durable thermoplastic rubber, these reusable earplugs feature a flanged, push-in design and are corded for secure and convenient use. They are suitable for various industrial and laboratory environments.
  • Noise Reduction Rating of 27 dB
  • Constructed from thermoplastic rubber
  • Flanged plug design
  • Push-in insertion method
  • Corded for secure placement
  • Non-metal detectable
  • Reusable for extended use
  • Universal size to fit most users
  • High visibility green color
Catalog No. 50-283-5483 Supplier MSC Supplier No. 18780726
Cordova earplugs provide effective hearing protection with a 27 dB Noise Reduction Rating. Constructed from durable thermoplastic rubber, these reusable earplugs feature a flanged, push-in design and are corded for secure and convenient use. They are suitable for various industrial and laboratory environments.
  • Noise Reduction Rating of 27 dB
  • Constructed from thermoplastic rubber
  • Flanged plug design
  • Push-in insertion method
  • Corded for secure placement
  • Non-metal detectable
  • Reusable for extended use
  • Universal size to fit most users
  • High visibility green color