Learn More
MSC Moldex Earplugs, corded, 33 dB NRR, foam, tapered
Catalog No. 502835042
Encompass
The Moldex SparkPlugs 6654 are corded disposable foam earplugs designed for noise reduction in various environments. They feature an extra-soft, low-pressure foam construction and a tapered shape for comfortable and easy insertion. With a Noise Reduction Rating (NRR) of 33 dB, these earplugs provide excellent hearing protection. They are 100% PVC-Free and independently tested to ANSI S3.19-1974 standards.
  • Extra-soft, low-pressure foam for comfort and fit.
  • Tapered shape for easy insertion and removal.
  • Designed to fit even very small ear canals.
  • Non-irritating smooth surface.
  • Exclusive colored streaks and swirls.
  • Independently tested to ANSI S3.19-1974.
  • 100% PVC-Free material.
  • 33 dB Noise Reduction Rating.
Catalog No. 50-283-5042 Supplier MSC Supplier No. 06506349
The Moldex SparkPlugs 6654 are corded disposable foam earplugs designed for noise reduction in various environments. They feature an extra-soft, low-pressure foam construction and a tapered shape for comfortable and easy insertion. With a Noise Reduction Rating (NRR) of 33 dB, these earplugs provide excellent hearing protection. They are 100% PVC-Free and independently tested to ANSI S3.19-1974 standards.
  • Extra-soft, low-pressure foam for comfort and fit.
  • Tapered shape for easy insertion and removal.
  • Designed to fit even very small ear canals.
  • Non-irritating smooth surface.
  • Exclusive colored streaks and swirls.
  • Independently tested to ANSI S3.19-1974.
  • 100% PVC-Free material.
  • 33 dB Noise Reduction Rating.