Learn More
MSC 3M Earplugs, 29 dB, Foam, Cylinder, Uncorded
Catalog No. 502835044
Encompass
These earplugs provide reliable hearing protection with a Noise Reduction Rating (NRR) of 29 dB. Made from soft foam with a cylindrical shape, they are designed for comfortable wear and effective noise attenuation. They are uncorded, reusable, and suitable for a universal fit.
  • Provides a Noise Reduction Rating of 29 dB
  • Constructed from foam material
  • Features a cylindrical plug shape
  • Designed for uncorded use
  • Reusable for extended use
  • Roll down insertion method
  • Universal size for a broad fit
  • Yellow color for visibility
  • Non-metal detectable
  • Part of the E-A-R Classic series
Catalog No. 50-283-5044 Supplier MSC Supplier No. 06528061
These earplugs provide reliable hearing protection with a Noise Reduction Rating (NRR) of 29 dB. Made from soft foam with a cylindrical shape, they are designed for comfortable wear and effective noise attenuation. They are uncorded, reusable, and suitable for a universal fit.
  • Provides a Noise Reduction Rating of 29 dB
  • Constructed from foam material
  • Features a cylindrical plug shape
  • Designed for uncorded use
  • Reusable for extended use
  • Roll down insertion method
  • Universal size for a broad fit
  • Yellow color for visibility
  • Non-metal detectable
  • Part of the E-A-R Classic series