Learn More
MSC 3M Earplugs, 32 dB, Foam, Corded, Metal-Detectable
Catalog No. 502835324
Encompass
These earplugs are designed for high-performance noise reduction, offering comfort for extended wear. They feature a soft, pliable foam construction, are corded for convenience, and are metal-detectable, making them suitable for sensitive environments like food processing where product contamination must be prevented.
  • Soft, pliable foam offers comfort for wearing over an extended time.
  • Implanted stainless steel ball bearing and metal particles aid in locating lost earplugs.
  • Regular size fits most earcanals.
  • Designed for food processing and other industries where it is critical to keep earplugs from contaminating products and processes.
  • Poly bag keeps each pair of earplugs clean before use.
Catalog No. 50-283-5324 Supplier MSC Supplier No. 85206571
These earplugs are designed for high-performance noise reduction, offering comfort for extended wear. They feature a soft, pliable foam construction, are corded for convenience, and are metal-detectable, making them suitable for sensitive environments like food processing where product contamination must be prevented.
  • Soft, pliable foam offers comfort for wearing over an extended time.
  • Implanted stainless steel ball bearing and metal particles aid in locating lost earplugs.
  • Regular size fits most earcanals.
  • Designed for food processing and other industries where it is critical to keep earplugs from contaminating products and processes.
  • Poly bag keeps each pair of earplugs clean before use.