Learn More
MSC 3M Earplugs, 25 dB NRR, Foam, Corded, Push-to-fit
Catalog No. 502835478
Encompass
3M E-A-R Express Pod Plugs earplugs feature advanced "no-roll", slow-recovery foam and paddle-style grips for quick, clean, and consistent placement. This unique design helps keep earplug tips clean even when hands are dirty. The slow recovery foam easily inserts into the ear canal, then seals it for comfortable hearing protection, offering a Noise Reduction Rating (NRR) of 25 dB.
  • Paddle-style grips provide quick, clean, and consistent placement.
  • No-roll design keeps earplugs and ears clean, even with dirty fingers or gloves.
  • Unique foam formula allows earplug insertion without rolling.
  • Slow recovery foam easily enters the ear canal, then seats and stays in place.
  • Test compatible with the E-A-Rfit Validation System.
  • Offers a Noise Reduction Rating (NRR) of 25 dB.
  • Reusable for extended use.
  • Features a push-to-fit insertion style.
Catalog No. 50-283-5478 Supplier MSC Supplier No. 67869750
3M E-A-R Express Pod Plugs earplugs feature advanced "no-roll", slow-recovery foam and paddle-style grips for quick, clean, and consistent placement. This unique design helps keep earplug tips clean even when hands are dirty. The slow recovery foam easily inserts into the ear canal, then seals it for comfortable hearing protection, offering a Noise Reduction Rating (NRR) of 25 dB.
  • Paddle-style grips provide quick, clean, and consistent placement.
  • No-roll design keeps earplugs and ears clean, even with dirty fingers or gloves.
  • Unique foam formula allows earplug insertion without rolling.
  • Slow recovery foam easily enters the ear canal, then seats and stays in place.
  • Test compatible with the E-A-Rfit Validation System.
  • Offers a Noise Reduction Rating (NRR) of 25 dB.
  • Reusable for extended use.
  • Features a push-to-fit insertion style.