Learn More
MSC 3M Earplugs, 29 dB NRR, Foam, Cylindrical, Corded
Catalog No. 502835039
Encompass
The 3M E-A-R Classic earplugs revolutionized hearing protection, offering a 29 dB Noise Reduction Rating. Made from a proprietary soft, slow-recovery foam, they conform to the ear canal for a comfortable and secure fit. These disposable, corded, cylindrical earplugs are bright yellow for easy visibility and convenient storage around the neck.
  • Soft, slow-recovery foam ensures comfort and aids in earplug fitting.
  • Moisture-resistant foam material.
  • Corded for convenient temporary storage around the neck.
  • Disposable for hygiene and convenience.
  • Provides 29 dB noise reduction rating.
  • Cylindrical shape for easy insertion and fit.
  • Compatible with personal fit validation systems.
  • Non-allergenic and non-flammable.
  • Electrically insulated.
  • Bright yellow color for quick visual compliance checks.
Catalog No. 50-283-5039 Supplier MSC Supplier No. 06528079
The 3M E-A-R Classic earplugs revolutionized hearing protection, offering a 29 dB Noise Reduction Rating. Made from a proprietary soft, slow-recovery foam, they conform to the ear canal for a comfortable and secure fit. These disposable, corded, cylindrical earplugs are bright yellow for easy visibility and convenient storage around the neck.
  • Soft, slow-recovery foam ensures comfort and aids in earplug fitting.
  • Moisture-resistant foam material.
  • Corded for convenient temporary storage around the neck.
  • Disposable for hygiene and convenience.
  • Provides 29 dB noise reduction rating.
  • Cylindrical shape for easy insertion and fit.
  • Compatible with personal fit validation systems.
  • Non-allergenic and non-flammable.
  • Electrically insulated.
  • Bright yellow color for quick visual compliance checks.