Learn More
MSC 3M Earplugs, 27 dB NRR, Foam, Flanged, Corded, Reusable
Catalog No. 502835165
Encompass
These earplugs offer a 27 dB Noise Reduction Rating, featuring a flanged, push-in design made from foam material. They are corded and reusable, providing effective hearing protection. Supplied in a pack of 100 pairs, they are suitable for various industrial and laboratory environments.
  • Noise reduction rating of 27 dB
  • Made from foam material
  • Flanged plug shape
  • Push-in insertion method
  • Corded for convenience
  • Reusable design
  • Universal size for broad applicability
  • Yellow plug color
  • Non-metal detectable
  • Vinyl cord material in blue
  • Conforms to CSA Class AL standards
  • Packaged in a cardboard box, 100 pairs
Catalog No. 50-283-5165 Supplier MSC Supplier No. 85684850
These earplugs offer a 27 dB Noise Reduction Rating, featuring a flanged, push-in design made from foam material. They are corded and reusable, providing effective hearing protection. Supplied in a pack of 100 pairs, they are suitable for various industrial and laboratory environments.
  • Noise reduction rating of 27 dB
  • Made from foam material
  • Flanged plug shape
  • Push-in insertion method
  • Corded for convenience
  • Reusable design
  • Universal size for broad applicability
  • Yellow plug color
  • Non-metal detectable
  • Vinyl cord material in blue
  • Conforms to CSA Class AL standards
  • Packaged in a cardboard box, 100 pairs