Learn More
Essendant LL-30 Laser Lite Single-Use Earplugs, Corded, 32NRR, Magenta/Yellow, 100 Pairs
Catalog No. 2789895
Encompass_Preferred
Vibrant coloring keeps your protection highly visible, enabling easy compliance sighting. Self-adjusting polyurethane foam expands upon insertion, ensuring a tight yet comfortable ear-canal seal. Smooth, soil-resistant closed-cell foam skin prevents dirt buildup. Contoured shape fits to a T.
Catalog No. 27-898-95 Supplier Essendant Supplier No. HOWLL30
Vibrant coloring keeps your protection highly visible, enabling easy compliance sighting. Self-adjusting polyurethane foam expands upon insertion, ensuring a tight yet comfortable ear-canal seal. Smooth, soil-resistant closed-cell foam skin prevents dirt buildup. Contoured shape fits to a T.