Learn More
MSC EARSOFT YELLOW NEONS
Catalog No. 5000782083
Encompass
3M E-A-Rsoft Yellow Neons Earplugs feature a tapered shape and smooth soft texture for a comfortable fit. Made of soft, slow-recovery foam, these earplugs aid easier insertion and offer 3M's maximum noise reduction for hearing protection, providing comfort for extended wear.
  • Tapered shape and smooth soft texture for fit and comfort
  • Slow-recovery foam designed for easier insertion
  • Compatible with the E-A-Rfit Validation System
  • Available with and without cords
  • Available in two sizes: regular and large
  • Noise Reduction Rating (NRR) of 33 dB
  • CSA Class AL
  • Extremely soft foam provides increasing comfort and wearability
  • Vibrant bright color
  • Supplied in polybag or dispenser refills
Catalog No. 50-007-82083 Supplier MSC Supplier No. 08353187
3M E-A-Rsoft Yellow Neons Earplugs feature a tapered shape and smooth soft texture for a comfortable fit. Made of soft, slow-recovery foam, these earplugs aid easier insertion and offer 3M's maximum noise reduction for hearing protection, providing comfort for extended wear.
  • Tapered shape and smooth soft texture for fit and comfort
  • Slow-recovery foam designed for easier insertion
  • Compatible with the E-A-Rfit Validation System
  • Available with and without cords
  • Available in two sizes: regular and large
  • Noise Reduction Rating (NRR) of 33 dB
  • CSA Class AL
  • Extremely soft foam provides increasing comfort and wearability
  • Vibrant bright color
  • Supplied in polybag or dispenser refills