Learn More
MSC EARSOFT YW NEON BLASTS
Catalog No. 5000676382
Encompass
These earplugs are designed to provide effective hearing protection in various environments. They feature a high noise reduction rating and a comfortable, reusable design, making them suitable for repeated use.
  • Reusable design
  • Uncorded attachment style
  • Bullet shape for comfortable fit
  • Noise reduction rating of 33 dB
  • Foam material construction
  • Roll down insertion method
  • Universal size
  • Non-metal detectable
  • High visibility yellow color
Catalog No. 50-006-76382 Supplier MSC Supplier No. 08353153
These earplugs are designed to provide effective hearing protection in various environments. They feature a high noise reduction rating and a comfortable, reusable design, making them suitable for repeated use.
  • Reusable design
  • Uncorded attachment style
  • Bullet shape for comfortable fit
  • Noise reduction rating of 33 dB
  • Foam material construction
  • Roll down insertion method
  • Universal size
  • Non-metal detectable
  • High visibility yellow color