Learn More
GENTECH SOLUTIONS LLC EARTH-Core 5G Soil Sampler
Catalog No. NC3728758
Encompass_Preferred
EliteChem EARTH-Core, an efficient, cost-effective, single use transfer tool designed for soil sampling in accordance with USE PA SW-846 Method 5035. The EARTH-Core is a compact, single-use and self contained device that streamlines soil sampling and facilitates the transfer of samples into containers for on-site chemical preservation. It delivers 5 gram samples, eliminating the need for any extra handle, making it simple and easy to use. No more complications with cut off syringes!
Catalog No. NC3728758 Supplier GENTECH SOLUTIONS LLC Supplier No. 1200100010
EliteChem EARTH-Core, an efficient, cost-effective, single use transfer tool designed for soil sampling in accordance with USE PA SW-846 Method 5035. The EARTH-Core is a compact, single-use and self contained device that streamlines soil sampling and facilitates the transfer of samples into containers for on-site chemical preservation. It delivers 5 gram samples, eliminating the need for any extra handle, making it simple and easy to use. No more complications with cut off syringes!