Learn More
Grainger Extech 4-Wire Earth Ground Resistance Tester, 2 to 2000 Ohm, CAT IV 300V, 1 ea
Catalog No. 502770261
Encompass_Preferred
The 4-Wire Earth Ground Resistance Tester Kit is designed for precise earth ground measurements across four ranges from 2 Ohm to 2,000 Ohm. It supports 2-wire, 3-wire, and 4-wire testing, making it a versatile tool for various field applications. The unit features a large dual-line LCD, automatic current and potential spike checking, and a rugged design suited for professional use.
  • Supports 2-wire, 3-wire, and 4-wire testing modes
  • Measures earth ground in four ranges from 2 Ohm to 2,000 Ohm
  • Includes automatic current and potential spike check for accuracy
  • Features autoranging, data hold, and automatic power off functions
  • Includes test leads with alligator clips, four auxiliary earth bars, a hard carrying case, and eight 1.5V AA batteries
  • Safety rated to CAT IV 300 V
  • Measurement range for AC earth voltage up to 300 VAC
Catalog No. 50-277-0261 Supplier Grainger Supplier No. 13X132
The 4-Wire Earth Ground Resistance Tester Kit is designed for precise earth ground measurements across four ranges from 2 Ohm to 2,000 Ohm. It supports 2-wire, 3-wire, and 4-wire testing, making it a versatile tool for various field applications. The unit features a large dual-line LCD, automatic current and potential spike checking, and a rugged design suited for professional use.
  • Supports 2-wire, 3-wire, and 4-wire testing modes
  • Measures earth ground in four ranges from 2 Ohm to 2,000 Ohm
  • Includes automatic current and potential spike check for accuracy
  • Features autoranging, data hold, and automatic power off functions
  • Includes test leads with alligator clips, four auxiliary earth bars, a hard carrying case, and eight 1.5V AA batteries
  • Safety rated to CAT IV 300 V
  • Measurement range for AC earth voltage up to 300 VAC