Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Earth Science Curriculum Mastery™ Posters
SCI_ED

Catalog No. S04389 Shop All NewPath Learning Products
Change view
Click to view available options
Product Type:
Earthquakes Poster
Rocks Poster
Understanding Weather Poster
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S04389 Rocks Poster
S04392 Understanding Weather Poster
S04391 Earthquakes Poster
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S04389 Supplier NewPath Learning Supplier No. 346502
Add to Cart
Add to Cart

Graphic overview of key earth science topics!

  • Provides a comprehensive look at a key, standards-based topic
  • Laminated
  • Grades 6-10

Minerals Poster:
What is a mineral? How do they form? How can they be identified? What are their uses?

Rocks Poster:
An illustrated look at metamorphic, sedimentary and igneous rocks as well as how they are formed!

Fossils Poster:
A fascinating look at fossils, their types, how they form and their use in studying the geologic past

Earthquakes Poster:
Provides a detailed look at faults, seismic waves and earthquakes as well as how they are measured

Understanding Weather Poster:
Features a comprehensive overview of weather; humidity and precipitation; the water cycle; clouds and their formation; and the use of weather maps

Order Info

Size: 23 x 35 in. (58.4 x 89cm)

Specifications

Product Type Rocks Poster
For Use With (Application) Provides an illustrated look at metamorphic, sedimentary and igneous rocks as well as how they formed
Certifications/Compliance Academic Standard for California, Florida, National, New York, Texas
Grade Level Intermediate - Middle School, High School
Grade 6 to 12
Subject Science
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.