Learn More
MSC Earth Sense Multi-Purpose Cleaner, 1 gal, Concentrated, pH Neutral, Green Seal
Catalog No. 502835638
Encompass
This multi-purpose pH neutral cleaner is environmentally responsible, formulated without phosphates, solvents, nonylphenol ethoxylate surfactants. It is readily biodegradable, free of VOCs, added fragrance, silicates, chelating agents, or caustics. Its water-based, concentrated, non-acid formula uses linear alcohol sulfonate surfactants for effective cleaning on finished floors, ceramic tile, painted surfaces, and other resilient and non-resilient surfaces.
  • High performance pH neutral cleaner
  • Environmentally responsible formulation
  • Free from phosphates, solvents, and nonylphenol ethoxylate surfactants
  • Readily biodegradable with no VOCs or added fragrance
  • Effective on a wide range of surfaces including finished floors, ceramic tile, and painted surfaces
  • Concentrated and water-based solution
  • Non-acid formula
Catalog No. 50-283-5638 Supplier MSC Supplier No. 75123133
This multi-purpose pH neutral cleaner is environmentally responsible, formulated without phosphates, solvents, nonylphenol ethoxylate surfactants. It is readily biodegradable, free of VOCs, added fragrance, silicates, chelating agents, or caustics. Its water-based, concentrated, non-acid formula uses linear alcohol sulfonate surfactants for effective cleaning on finished floors, ceramic tile, painted surfaces, and other resilient and non-resilient surfaces.
  • High performance pH neutral cleaner
  • Environmentally responsible formulation
  • Free from phosphates, solvents, and nonylphenol ethoxylate surfactants
  • Readily biodegradable with no VOCs or added fragrance
  • Effective on a wide range of surfaces including finished floors, ceramic tile, and painted surfaces
  • Concentrated and water-based solution
  • Non-acid formula