Learn More
MSC Spray Nine Bio-Based Cleaner, Industrial Strength, Degreaser, 55 Gal
Catalog No. 502866236
Encompass
This environmentally friendly, biodegradable industrial strength cleaner/degreaser is a bio-based safe alternative to solvent and caustic cleaning products. It easily removes dirt, grease, carbon, oil, adhesives, and soot.
  • Industrial strength cleaner and degreaser
  • Bio-based safe alternative to solvent and caustic cleaning products
  • Corrosion inhibiting formula for use in traditional and heated parts washers (up to 180F)
  • Biodegradable, water soluble solution
Catalog No. 50-286-6236 Supplier MSC Supplier No. 76843218
This environmentally friendly, biodegradable industrial strength cleaner/degreaser is a bio-based safe alternative to solvent and caustic cleaning products. It easily removes dirt, grease, carbon, oil, adhesives, and soot.
  • Industrial strength cleaner and degreaser
  • Bio-based safe alternative to solvent and caustic cleaning products
  • Corrosion inhibiting formula for use in traditional and heated parts washers (up to 180F)
  • Biodegradable, water soluble solution