Learn More
Mopec EasiCollect+ Devices (EC+)
SDP
Catalog No. 22445974 Shop All Mopec Products
Change view
Click to view available options
Quantity:
50/Pk.
Catalog No. Quantity
22-445-974 50/Pk.
1 options
Catalog No. 22-445-974 Supplier Mopec Supplier No. FS240
Add to Cart
Add to Cart
Designed to improve collection efficiency and reliability while stabilizing DNA for Long-term storage.
  • Includes a card, swab, and integrated sample collection device.
  • Incorporates Whatman™ FTA™ card technology for long-term sample stabilization and supports consistent biological sample collection.
  • Provides a 98% First-pass success rate in field trials and laboratory models
  • Helps simplify and improve consistency in sample collection.
Specifications
Product Type Cell Collection Device
Color Blue, White
Quantity 50/Pk.