Learn More
ValueMax™ Easy-Breathe™ Lab Coats, Ceil Blue
Catalog No. 23900754
Change view
Click to view available options
Size:
4X-Large
5X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
23-900-754 4X-Large
23-900-755 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-900-754 Supplier ValueMax™ Supplier No. 3560CB4XL
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
This item is not returnable. View return policy
Great combination of fluid resistance and breathability
  • Silky soft 'SMS' fabric which meets AAMI level 2 fluid resistant standards
  • Anti-static treated and clinging-free
  • Knee length lab coats with snap front and three pockets
  • Knitted cuffs and collar
  • Economical
  • Color: Ceil Blue
Specifications
Product Type General Purpose Lab Coat
Size 4X-Large
Gender Unisex
Material SMS
Color Ceil Blue
Cuff Style Knit
Collar Type Knit