Learn More
ValueMax™ Easy-Breathe™ Lab Coats, Ceil Blue
Catalog No. 23900755
Change view
Click to view available options
Size:
4X-Large
5X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
23-900-755 5X-Large
23-900-754 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-900-755 Supplier ValueMax™ Supplier No. 3560CB5XL
This item is not returnable. View return policy
Great combination of fluid resistance and breathability
  • Silky soft 'SMS' fabric which meets AAMI level 2 fluid resistant standards
  • Anti-static treated and clinging-free
  • Knee length lab coats with snap front and three pockets
  • Knitted cuffs and collar
  • Economical
  • Color: Ceil Blue
Specifications
Product Type General Purpose Lab Coat
Size 5X-Large
Gender Unisex
Material SMS
Color Ceil Blue
Cuff Style Knit
Collar Type Knit