Learn More
Perkin Elmer US LLC Easy Diff Small Sample Cup - 10 mm, Qty. 2
Catalog No. 502113236
Encompass_Preferred
The EasyDiff Diffuse Reflectance Accessory has pre-aligned optical components and micrometer-controlled sample positioning for reproducible, high quality data analysis of powders, solids, minerals, and inorganic solids. The EasyDiff accessory is compatible with Spectrum Two and Spectrum 3/Frontier spectrometers.
Catalog No. 50-211-3236 Supplier Perkin Elmer US LLC Supplier No. L1272346
The EasyDiff Diffuse Reflectance Accessory has pre-aligned optical components and micrometer-controlled sample positioning for reproducible, high quality data analysis of powders, solids, minerals, and inorganic solids. The EasyDiff accessory is compatible with Spectrum Two and Spectrum 3/Frontier spectrometers.