Learn More
Research Products International Corp Easy Dip Staining Jar, Pink, 6 per Package
SDP
Catalog No. 501977960
Encompass
Unique slide staining system features a square staining jar and a vertical slide rack. Both components are made from heavy duty acetal polymer resistant to alcohol and Xylene. Staining jar has a wide stable base and a recessed interior cavity with a reduced volume capacity of 80ml. Slide staining rack holds up to 12 slides with dimensions of 25 x 75mm, 25x76mm or 1" x 3". Slides are held vertically for easy insertion and removal. Rack is designed for quick drying of slides in a microwave oven or at temperatures up to 120°C. Jars available in five different colors for sample identification.
Jars available in five different colors for sample identification. Each jar is supplied with clip for quick and easy connection of staining jars.
Slide staining rack is sold separately.
Catalog No. 50-197-7960 Supplier Research Products International Corp Supplier No. 247835P
Unique slide staining system features a square staining jar and a vertical slide rack. Both components are made from heavy duty acetal polymer resistant to alcohol and Xylene. Staining jar has a wide stable base and a recessed interior cavity with a reduced volume capacity of 80ml. Slide staining rack holds up to 12 slides with dimensions of 25 x 75mm, 25x76mm or 1" x 3". Slides are held vertically for easy insertion and removal. Rack is designed for quick drying of slides in a microwave oven or at temperatures up to 120°C. Jars available in five different colors for sample identification.
Jars available in five different colors for sample identification. Each jar is supplied with clip for quick and easy connection of staining jars.
Slide staining rack is sold separately.