Learn More
Newcomer Supply EasyDip Slide Staining Kit, 5 Jars (One of each color: Blue, Green, Pink, Whitek, & Yellow) & 1 Rack
SDP
Catalog No. NC1649459
Encompass
The EasyDip Slide Staining Kit contains 5 EasyDip Slide Staining Jars (one of each color) and 1 EasyDip Slide Staining Rack. The EasyDip Slide Staining Jar is made of acetal polymer and has an attached lid. The jar uses just 80ml reagent for 12 slides and is resistant to most staining agents including alcohol and xylene. Temperature range: -80°C to +30°C. Dimensions: 2 1/2" x 3" x 3 5/8" (H). The jars can be used individually or linked together and can be used with the EasyDip Slide Staining Rack.
Catalog No. NC1649459 Supplier Newcomer Supply Supplier No. 5300KIT
The EasyDip Slide Staining Kit contains 5 EasyDip Slide Staining Jars (one of each color) and 1 EasyDip Slide Staining Rack. The EasyDip Slide Staining Jar is made of acetal polymer and has an attached lid. The jar uses just 80ml reagent for 12 slides and is resistant to most staining agents including alcohol and xylene. Temperature range: -80°C to +30°C. Dimensions: 2 1/2" x 3" x 3 5/8" (H). The jars can be used individually or linked together and can be used with the EasyDip Slide Staining Rack.