Learn More
Electron Microscopy Sciences Easy Dip™ Kit (Slide Staining Jar and Rack), White
SDP
Catalog No. NC0263028
Encompass_Preferred
The EasyDip Slide Staining system has a square staining jar and a 12-position vertical slide staining rack. Jars can be loosely joined to each other laterally.
The Staining Jar is made of acetal plastic and resists attacks from most staining reagents including alcohol and xylenes but not phenols. The wide stable base offers stability while the inside is recessed, allowing for a smaller reagent volume of only 80 ml. Ideal for frozen sections, small jobs, and special processes.
The EasyDip Slide Staining Rack will hold up to 12 slides of 25x75 mm with a thickness of 1.0 to 1.2 mm. These racks are safe up to 120 deg C. The lid completely covers the EasyDip Slide Staining Jar to minimize spill and evaporation. Handle is permanently attached to the rack for easy insertion or removal of the slide. A square staining jar and a 12-position vertical slide staining rack.
Color: White
Catalog No. NC0263028 Supplier Electron Microscopy Sciences Supplier No. 7138801
The EasyDip Slide Staining system has a square staining jar and a 12-position vertical slide staining rack. Jars can be loosely joined to each other laterally.
The Staining Jar is made of acetal plastic and resists attacks from most staining reagents including alcohol and xylenes but not phenols. The wide stable base offers stability while the inside is recessed, allowing for a smaller reagent volume of only 80 ml. Ideal for frozen sections, small jobs, and special processes.
The EasyDip Slide Staining Rack will hold up to 12 slides of 25x75 mm with a thickness of 1.0 to 1.2 mm. These racks are safe up to 120 deg C. The lid completely covers the EasyDip Slide Staining Jar to minimize spill and evaporation. Handle is permanently attached to the rack for easy insertion or removal of the slide. A square staining jar and a 12-position vertical slide staining rack.
Color: White