Learn More
Newcomer Supply EasyDip Slide Staining Rack
SDP
Catalog No. NC0634808
Encompass
The EasyDip Slide Staining Rack is made of acetal polymer and holds up to 12 slides vertically. The lid completely covers the EasyDip Slide Staining Jar to minimize spill and evaporation. The handle is permanently attached to the rack for easy insertion and removal of slides without your fingers touching the solution. Do not use in microwave. Avoid using in picric acid and phenol for prolonged periods of time. Temperature range: -80°C to +30°C. Dimensions: 2 1/4" x 2 1/2" x 3 3/4" (H). Available in dark gray only. Designed to be used with the EasyDip Slide Staining Jar.
Catalog No. NC0634808 Supplier Newcomer Supply Supplier No. 5300RK
The EasyDip Slide Staining Rack is made of acetal polymer and holds up to 12 slides vertically. The lid completely covers the EasyDip Slide Staining Jar to minimize spill and evaporation. The handle is permanently attached to the rack for easy insertion and removal of slides without your fingers touching the solution. Do not use in microwave. Avoid using in picric acid and phenol for prolonged periods of time. Temperature range: -80°C to +30°C. Dimensions: 2 1/4" x 2 1/2" x 3 3/4" (H). Available in dark gray only. Designed to be used with the EasyDip Slide Staining Jar.