Learn More
Agilent Technologies 5100 DV ICP-OES Torches
Catalog No. NC1558765
Encompass
Easy-fit torch, demountable, for high solids, with 2.4 mm id injector and base assembly, with replaceable outer and intermediate tubes, for use with Agilent 5100 Synchronous Vertical Dual View/Vertical Dual View ICP-OES in axial and radial views.
Catalog No. NC1558765 Supplier Agilent Technologies Supplier No. G801060232
Easy-fit torch, demountable, for high solids, with 2.4 mm id injector and base assembly, with replaceable outer and intermediate tubes, for use with Agilent 5100 Synchronous Vertical Dual View/Vertical Dual View ICP-OES in axial and radial views.