Learn More
Vernier™ Easy to Go Adapter
SDPSCI_ED
Catalog No. S16203ND Shop All Vernier Software Products
Change view
Click to view available options
For Use With (Equipment):
EasyTemp and EasyLink
Catalog No. For Use With (Equipment)
S16203ND EasyTemp and EasyLink
1 options
Catalog No. S16203ND Supplier Vernier™ Supplier No. MINIUSB
Add to Cart
Add to Cart
Includes
Easy to Go Adapter
This adapter allows an EasyTemp or EasyLink to be connected to a computer for data collection using or Logger Lite software. The adapter has a USB Mini-A receptacle that connects to the EasyTemp or EasyLink and a standard-A plug that connects to the computer. It can also be used with the USB cable that comes with the CBR 2.
  • Connector 1: USB Mini Type A Receptacle
  • Connector 2: USB Standard Type A Plug
  • The adapter has a USB Mini-A receptacle that connects to the EasyTemp or EasyLink and a standard-A plug that connects to the computer.
  • It can also be used with the USB cable that comes with the CBR 2.
Warranty and Services
5-year limited warranty
Specifications
For Use With (Equipment) EasyTemp and EasyLink
For Use With (Application) Educational use only