Learn More
Bellco Glass, Inc. 40 Place Easy-Grip Workstation Rack for CF Cryogenic Vial, Non-sterile
SDP
Catalog No. 502329556
Encompass_Preferred
Easy-Grip Workstation Rack has individual positions which feature ribs to interlock with self-standing vials for easy one hand opening, features four gripping feet for increased stability and reduced slipping, and has low-profile design for maximum storage efficiency\n
    \n
  • Case Quantity: 2
  • \n
Catalog No. 50-232-9556 Supplier Bellco Glass, Inc. Supplier No. 229941000
Easy-Grip Workstation Rack has individual positions which feature ribs to interlock with self-standing vials for easy one hand opening, features four gripping feet for increased stability and reduced slipping, and has low-profile design for maximum storage efficiency\n
    \n
  • Case Quantity: 2
  • \n