Learn More
Anton Paar Easy handling with workflow support enables convenient digestion. Multiple safety systems ensure reliability. Offers PDC benefits at a competitive price. Smart solution for most samples, with optional external cooling for added cost savings.
Catalog No. 5000580494
Encompass_Preferred
High temperature and pressure for outstanding digestion resultsSoftware-guided procedures make the instrument easy to useIntegrated cooling system for a space-saving deviceLarge selection of vials and vessels for all analytical problemsVisual and digital status updates
Catalog No. 50-005-80494 Supplier Anton Paar Supplier No. 266260
Please contact your sales representative for a quote or further ordering instructions.
High temperature and pressure for outstanding digestion resultsSoftware-guided procedures make the instrument easy to useIntegrated cooling system for a space-saving deviceLarge selection of vials and vessels for all analytical problemsVisual and digital status updates