Learn More
MSC PEC Tools Steel Rigid Rule, 18 in., 5R Graduation, 1 1/8 in. Wide, 1 ea.
Catalog No. 5000513208
Encompass
This rigid steel rule is designed for precision dimensional measurement applications. Constructed from high-quality hardened and tempered steel, it features a durable black chrome finish that provides rust resistance and facilitates clear reading in varying light conditions. The tool is manufactured to meet demanding shop standards with precision-ground edges and clearly etched graduations.
  • Features 5R graduation style including 1/32 in., 1/64 in., 1/10 in., and 1/100 in. increments
  • Constructed from hardened and tempered steel for durability
  • Black chrome finish for enhanced readability and corrosion resistance
  • Precision ground flat and straight edges
  • Made in the USA
Catalog No. 50-005-13208 Supplier MSC Supplier No. 06511448
This rigid steel rule is designed for precision dimensional measurement applications. Constructed from high-quality hardened and tempered steel, it features a durable black chrome finish that provides rust resistance and facilitates clear reading in varying light conditions. The tool is manufactured to meet demanding shop standards with precision-ground edges and clearly etched graduations.
  • Features 5R graduation style including 1/32 in., 1/64 in., 1/10 in., and 1/100 in. increments
  • Constructed from hardened and tempered steel for durability
  • Black chrome finish for enhanced readability and corrosion resistance
  • Precision ground flat and straight edges
  • Made in the USA