Learn More
METTLER TOLEDO Mettler Toledo 100 mL Glass Cover Set, Chemically Resistant, 6 Ports
Catalog No. NC0754099 Shop All METTLER TOLEDO Products
Encompass
The METTLER TOLEDO 100 mL Glass Cover Set is a chemically-resistant accessory designed for EasyMax reactor systems, enabling optimal reaction monitoring through a 6-port design. Its transparent construction facilitates easy observation and supports multiple analytical probes, making it ideal for chemical and pharmaceutical research. The set is ready-to-use and includes essential components, offering a combination of functionality and convenience for lab experiments.
Catalog No. NC0754099 Supplier METTLER TOLEDO Supplier No. 51161709
The METTLER TOLEDO 100 mL Glass Cover Set is a chemically-resistant accessory designed for EasyMax reactor systems, enabling optimal reaction monitoring through a 6-port design. Its transparent construction facilitates easy observation and supports multiple analytical probes, making it ideal for chemical and pharmaceutical research. The set is ready-to-use and includes essential components, offering a combination of functionality and convenience for lab experiments.